missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human CD58 / LFA-3 Protein, His Tag
353.08 CHF - 2244.59 CHF
Spezifikation
Zugriffsnummer | AAH05930 |
---|---|
Molekulargewicht | 22.3 kDa |
Quelle | Human CD58, His Tag (LF3-H5225) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession # AAH05930). |
Immunogen | Phe 29 - Arg 215 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15827247
|
ACROBiosystems
LF3-H5225-1MG |
1 mg |
2244.59 CHF
1 Milligramm |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15817247
|
ACROBiosystems
LF3-H5225-100UG |
100 μg |
353.08 CHF
100 Mikrogramm |
Verfügbar ab: 07-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
CD antigen CD58 is also known as Lymphocyte function-associated antigen 3 (LFA-3). CD58 is a cell adhesion molecule expressed on Antigen Presenting Cells (APC), particularly macrophages. CD58 /LFA3 is ligand of the T-lymphocyte CD2 glycoprotein, which binds to CD2 (LFA-2) [3] on T cells and is important in strengthening the adhesion between the T cells and Professional Antigen Presenting Cells. In addition, the LFA-3/CD2 interaction may prime response by both the CD2+ and LFA-3+ cells. Polymorphisms in the CD58 gene are associated with increased risk for multiple sclerosis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa. The protein migrates as 30-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH05930 | |
Human CD58, His Tag (LF3-H5225) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession # AAH05930). | |
-20°C | |
CD58 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
22.3 kDa | |
Phe 29 - Arg 215 | |
CD58,LFA3,Ag3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |