Learn More
ACROBiosystems Human CD58 / LFA-3 Protein, His Tag
Marke: ACROBiosystems LF3-H5225-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
CD antigen CD58 is also known as Lymphocyte function-associated antigen 3 (LFA-3). CD58 is a cell adhesion molecule expressed on Antigen Presenting Cells (APC), particularly macrophages. CD58 /LFA3 is ligand of the T-lymphocyte CD2 glycoprotein, which binds to CD2 (LFA-2) [3] on T cells and is important in strengthening the adhesion between the T cells and Professional Antigen Presenting Cells. In addition, the LFA-3/CD2 interaction may prime response by both the CD2+ and LFA-3+ cells. Polymorphisms in the CD58 gene are associated with increased risk for multiple sclerosis.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.3 kDa. The protein migrates as 30-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH05930 | |
1 mg | |
Phe 29 - Arg 215 | |
CD58,LFA3,Ag3 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
22.3 kDa | |
Human CD58, His Tag (LF3-H5225) is expressed from human 293 cells (HEK293). It contains AA Phe 29 - Arg 215 (Accession # AAH05930). | |
-20°C | |
CD58 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |