Learn More
ACROBiosystems Human CD44 Protein, Fc Tag
HPLC-verified
315.25 CHF - 2320.25 CHF
Spezifikation
Zugriffsnummer | AAH04372 |
---|---|
Molekulargewicht | 48.7 kDa |
Quelle | Human CD44, Fc Tag (HPLC-verified) (PG1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Pro 220 (Accession # AAH04372). |
Immunogen | Gln 21 - Pro 220 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15817537
|
ACROBiosystems
PG1-H5255-100UG |
100 μg |
315.25 CHF
100 Mikrogramm |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15827537
|
ACROBiosystems
PG1-H5255-1MG |
1 mg |
2320.25 CHF
1 Milligramm |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
CD44 antigen is a cell-surface glycoprotein involved in cell–cell interactions, cell adhesion and migration. CD44 is expressed in a large number of mammalian cell types. CD44 is a receptor for hyaluronic acid and can also interact with other ligands, such as osteopontin, collagens, and matrix metalloproteinases (MMPs). CD44 function is controlled by its posttranslational modifications. One critical modification involves discrete sialofucosylations rendering the selectin-binding glycoform of CD44 called HCELL (for Hematopoietic Cell E-selectin/L-selectin Ligand). CD44 participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing, hematopoiesis, and tumor metastasis. Transcripts for this gene undergo complex alternative splicing that results in many functionally distinct isoforms.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.7 kDa. The protein migrates as 55-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH04372 | |
Human CD44, Fc Tag (HPLC-verified) (PG1-H5255) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Pro 220 (Accession # AAH04372). | |
-20°C | |
CD44 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
48.7 kDa | |
Gln 21 - Pro 220 | |
CD44,CDW44,CSPG8,ECMR-III,HCELL,HUTCH-I,IN,LHR,MC56,MDU2,MDU3,MIC4,Pgp1,Epican | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |