missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human CD4 Protein, Fc Tag
MALS verified
416.13 CHF - 1828.46 CHF
Spezifikation
Zugriffsnummer | AAH25782 |
---|---|
Molekulargewicht | 67.5 kDa |
Quelle | Human CD4, Fc Tag (MALS verified) (CD4-H5259) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # AAH25782). |
Immunogen | Lys 26 - Pro 396 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15892795
|
ACROBiosystems
CD4-H5259-200UG |
200 μg |
416.13 CHF
200 Mikrogramm |
Verfügbar ab: 02-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15826197
|
ACROBiosystems
CD4-H5259-1MG |
1 mg |
1828.46 CHF
1 Milligramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
Beschreibung
Cluster of Differentiation 4 (CD4) is also known as T-cell surface antigen T4/Leu-3 (LEU-3) and CD4mut, is a single-pass type I membrane glycoprotein,and is a member of the immunoglobulin superfamily. CD4 expressed on the surface of T helper cells, monocytes, macrophages, and dendritic cells. It has four immunoglobulin domains (D1 to D4) that are exposed on the extracellular surface of the cell: D1 and D3 resemble immunoglobulin variable (IgV) domains. D2 and D4 resemble immunoglobulin constant (IgC) domains. CD4 is a co-receptor that assists the T cell receptor (TCR) with an antigen-presenting cell. Using its portion that resides inside the T cell, CD4 amplifies the signal generated by the TCR by recruiting an enzyme, known as the tyrosine kinase lck, which is essential for activating many molecules involved in the signaling cascade of an activated T cell. CD4 also interacts directly with MHC class II molecules on the surface of the antigen-presenting cell using its extracellular domain. The extracellular domain adopts an immunoglobulin-like beta-sandwich with seven strands in 2 beta sheets, in a Greek key topology. CD4 has also been shown to interact with SPG21, Lck and Protein unc-119 homolog. CD4 is a primary receptor used by HIV-1 to gain entry into host T cells. HIV infection leads to a progressive reduction of the number of T cells possessing CD4 receptors. Therefore, medical professionals refer to the CD4 count to decide when to begin treatment for HIV-infected patients. This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 67.5 kDa. The protein migrates as 70-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH25782 | |
Human CD4, Fc Tag (MALS verified) (CD4-H5259) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # AAH25782). | |
-20°C | |
CD4 | |
Human | |
Tris, Glycine and NaCl, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |
67.5 kDa | |
Lys 26 - Pro 396 | |
CD4,CD4mut,LEU3 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |