Learn More
ACROBiosystems Human CD3 epsilon&CD3 gamma Heterodimer Protein, Fc,His Tag&Fc,Flag Tag
MALS verified
Marke: ACROBiosystems CDG-H52W9-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
T-cell surface glycoprotein CD3 delta & CD3 gamma chain, also known as CD3D & CD3G or CD3D&CD3G respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
Human CD3E&CD3G Heterodimer Protein, Fc,His Tag&Fc,Flag Tag is produced by co-expression of CD3E and CD3G, has a calculated MW of 39.2 kDa (CD3E) and 38.6 kDa (CD3G). Subunit CD3E is fused with a human IgG1 Fc tag at the C-terminus, followed by a polyhistSpezifikation
NP_000724.1 (CD3E) and AAI13831.1 (CD3G) | |
1 mg | |
Asp 23 - Asp 126 (CD3E) and Gln 23 - Ser 116 (CD3G) | |
CD3 epsilon and CD3 gamma,CD3E and CD3G | |
Unconjugated | |
His Tag and Fc Tag and Flag Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
39.2 kDa (CD3E) and 38.6 kDa (CD3G) | |
Human CD3E&CD3G; Heterodimer Protein, Fc,His Tag&Fc;,Flag Tag(MALS verified) (CDG-H52W9) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Asp 126 (CD3E) and Gln 23 - Ser 116 (CD3G) (Accession # P07766-1 (CD3E) and P09693-1 (CD3G)). | |
-20°C | |
CD3E and CD3G | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |