Learn More
R&D Systems™ Human CD23/Fc epsilon RII (NP_001993) VersaClone cDNA
Description
This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.
Order Info
Ships at ambient temperature
Specifications
Specifications
| Gen | CD23/Fc epsilon RII |
| Zugriffsnummer | NP_001993 |
| Konzentration | 10 μg at 0.2 μg/μL |
| Zusammensetzung | cDNA is provided in 10mM Tris-Cl, pH 8.180 |
| Einsatztyp | 978bp |
| Gen-ID (Entrez) | 2208 |
| Gen-Alias | BLAST-2, CD23, CD23A, CD23CD23 antigen, CLEC4J, CLEC4JC-type lectin domain family 4 member J, C-type lectin domain family 4, member J, Fc epsilon RII, Fc fragment of IgE, low affinity II, receptor for (CD23), FCE2Fc fragment of IgE, low affinity II, receptor for (CD23A), fc-epsilon-RII, FCER2, Fcer2a, FceRII, IGEBF, Immunoglobulin E-binding factor, low affinity immunoglobulin epsilon Fc receptor, Ly-42, Lymphocyte IgE receptor |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Spezies | Human |
| Produktlinie | R&D Systems |
| Show More |
For Reasearch Use Only. Not for use in Humans.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.