Learn More
ACROBiosystems Human CD200 / OX-2 Protein, Fc Tag
Marke: ACROBiosystems OX2-H5251-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
CD200 is also known as OX-2 membrane glycoprotein (OX-2), is a type-1 membrane glycoprotein, which contains two immunoglobulin domains (1 Ig-like C2-type (immunoglobulin-like) domain and Ig-like V-type (immunoglobulin-like) domain), and thus belongs to the immunoglobulin superfamily. CD200 / OX-2 is widely expressed in multiple cell types. CD200 interacts with a structurally related receptor (CD200R) expressed mainly on myeloid cells and is involved in regulation of macrophage and mast cell function. OX-2 / CD200 and CD200R associate via their respective N-terminal Ig-like domains. CD200 also plays an important role in prevention of graft rejection, autoimmune diseases and spontaneous abortion.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 48.6 kDa. The protein migrates as 55-68 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH22522 | |
1 mg | |
Gln 31 - Gly 232 | |
CD200,MOX1,MOX2,MRC,OX-2,My033 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
48.6 kDa | |
Human CD200, Fc Tag (O x 2-H5251) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # AAH22522). | |
-20°C | |
CD200 | |
Human | |
50 mM Tris, 100 mM Glycine, pH7.5 | |
HEK293 | |
Bio-Markers and CD Antigens |