missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human CD163 Protein, His Tag

Marke:  ACROBiosystems CD3-H52E8-1MG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15826177

  • 2533.22 CHF / 1 Milligramm


Verfügbar ab
26-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Scavenger receptor cysteine-rich type 1 protein M130 (CD163) is also known as hemoglobin scavenger receptor, which is a scavenger receptor for the hemoglobin-haptoglobin complex. CD163 has also been shown to mark cells of monocyte/macrophage lineage. A soluble form of the receptor exists in plasma, commonly denoted sCD163. sCD163 is generated by ectodomain shedding of the membrane bound receptor. sCD163 is upregulated in a large range of inflammatory diseases including liver cirrhosis, type 2 diabetes, macrophage activation syndrome, Gaucher's disease, sepsis, HIV infection, rheumatoid arthritis and Hodgkin Lymphoma.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 110.4 kDa. The protein migrates as 135-140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

Q86VB7
HEK293
Human
110.4 kDa
1 mg
Ser 42 - Ser 1045
CD163,M130
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Bio-Markers and CD Antigens
Powder
>95% as determined by SDS-PAGE
Human CD163, His Tag (CD3-H52E8) is expressed from human 293 cells (HEK293). It contains AA Ser 42 - Ser 1045 (Accession # Q86VB7).
-20°C
CD163
His Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation