missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD163 Biotinylated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marke: R&D Systems BAM16072
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD163 Monoclonal specifically detects CD163 in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).
Spezifikation
| CD163 | |
| Monoclonal | |
| LYOPH | |
| ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL | |
| CAA80543 | |
| CD163 | |
| Mouse myeloma cell line NS0-derived recombinant human CD163 Gly41-Ser1045 Accession # CAA80543 | |
| 250 μg | |
| Primary | |
| Detects human CD163 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant mouse (rm) CD163, recombinant human (rh) TWEAK, rhCD6, rmTWEAK, or rmCD6 is observed. | |
| Human | |
| Purified |
| ELISA | |
| 215901 | |
| Biotin | |
| Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative | |
| CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9332 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur