Learn More
Human CD163 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 12 publications
Marke: R&D Systems AF1607-SP
219.30 CHF gültig bis 2025-12-16
BESTER Aktionspreis! Benutzen Sie den Promotions-Code "24090" um Ihren Promotions-Preis zu erhalten.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CD163 Polyclonal specifically detects CD163 in Human samples. It is validated for Western Blot, Simple Western.
Spezifikation
| CD163 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD_antigen: CD163, CD163, CD163 molecule, GHI/61, HbSR, Hemoglobin scavenger receptor, M130, macrophage-associated antigen, MM130, RM3/1, SCARI1, scavenger receptor cysteine-rich type 1 protein M130, sCD163, Soluble CD163 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9332 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| CAA80543 | |
| CD163 | |
| Mouse myeloma cell line NS0-derived recombinant human CD163 (R&D Systems, Catalog # 1607-CD) Gly41-Ser1045 Accession # CAA80543 | |
| 25 μg | |
| Primary | |
| Detects human CD163 in direct ELISAs and Western blots. | |
| Human | |
| IgG |