Learn More
ACROBiosystems Human CD14 Protein, His Tag
327.86 CHF - 2748.99 CHF
Spezifikation
Zugriffsnummer | AAH10507 |
---|---|
Molekulargewicht | 35.9 kDa |
Quelle | Human CD14, His Tag (CD4-H5228) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Met 344 (Accession # AAH10507). |
Immunogen | Thr 20 - Met 344 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15826187
|
ACROBiosystems
CD4-H5228-100UG |
100 μg |
327.86 CHF
100 Mikrogramm |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15836187
|
ACROBiosystems
CD4-H5228-1MG |
1 mg |
2748.99 CHF
1 Milligramm |
Verfügbar ab: 17-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Cluster of differentiation 14 (CD14), is a cell surface glycoprotein, and is a is a component of the innate immune system. CD14 is a myelomonocytic differentiation antigen preferentially expressed on monocytes, macrophages, and activated granulocytes. CD14 exists in two forms. Either it is anchored into the membrane by a glycosylphosphatidylinositol tail (mCD14) or it appears in a soluble form (sCD14). Soluble CD14 either appears after shedding of mCD14 (48 kDa) or is directly secreted from intracellular vesicles (56 kDa). CD14 acts as a co-receptor (along with the Toll-like receptor TLR 4 and MD-2) for the detection of bacterial lipopolysaccharide (LPS). CD14 can bind LPS only in the presence of lipopolysaccharide-binding protein (LBP). CD14 has been proposed to be involved in various biological processes, including transportation of other lipids, cell-cell interaction during different immune responses, as well as recognition of apoptotic cells. Although LPS is considered its main ligand, CD14 also recognizes other pathogen-associated molecular patterns. CD14+ cells are monocytes that can differentiate into a host of different cells. CD14 has been shown to interact with Lipopolysaccharide-binding protein.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 35.9 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH10507 | |
Human CD14, His Tag (CD4-H5228) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Met 344 (Accession # AAH10507). | |
-20°C | |
CD14 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
35.9 kDa | |
Thr 20 - Met 344 | |
CD14 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>98% as determined by SDS-PAGE |