missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CCL19/MIP-3 beta Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Marke: R&D Systems MAB361-100
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
CCL19/MIP-3 beta Monoclonal specifically detects CCL19/MIP-3 beta in Human samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry.
Spezifikation
| CCL19/MIP-3 beta | |
| Monoclonal | |
| Unconjugated | |
| Supplied as a 0.2 μm filtered solution in Tris and Acetic Acid. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta chemokine exodus-3, Beta-chemokine exodus-3, CC chemokine ligand 19, C-C motif chemokine 19, chemokine (C-C motif) ligand 19, CKb11, EBI1-ligand chemokine, ELC, ELCMIP-3-beta, Epstein-Barr virus-induced molecule 1 ligand chemokine, exodus-3, Macrophage inflammatory protein 3 beta, macrophage inflammatory protein 3-beta, MGC34433, MIP3 beta, MIP-3 beta, MIP-3b, MIP3BCK beta-11, SCYA19EBI1 ligand chemokine, small inducible cytokine subfamily A (Cys-Cys), member 19, Small-inducible cytokine A19 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 6363 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C, as supplied. 1 month, 2 to 8 °C under sterile conditions after opening. 6 months, -20 to -70 °C under sterile conditions after opening. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 54909 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
| Q99731.1 | |
| CCL19 | |
| E. coli-derived recombinant human CCL19/MIP-3 beta Gly22-Ser98 Accession # Q99731.1 | |
| 100 μg | |
| Primary | |
| Detects human CCL19/MIP-3 beta in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CCL1, 2, 3, 4, 5, 7, 8, 11, 13, 14, 15, 16, 17, 18, 20, 21, 22, 23, 24, 25, recombinant mouse CCL2, 3, 4, 5, 6, 7, 9, 11, 12, 21, 22, or 25 is observed. | |
| Human | |
| Ascites |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur