missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Cathepsin K ELISA Kit
For the quantitative detection of Human Cathepsin K
Marke: Invitrogen™ EEL033
Additional Details : Gewicht : 0.40000kg
Beschreibung
The protein encoded by this gene is a lysosomal cysteine proteinase involved in bone remodeling and resorption. This protein, which is a member of the peptidase C1 protein family, is predominantly expressed in osteoclasts. However, the encoded protein is also expressed in a significant fraction of human breast cancers, where it could contribute to tumor invasiveness. Mutations in this gene are the cause of pycnodysostosis, an autosomal recessive disease characterized by osteosclerosis and short stature. This gene may be subject to RNA editing.Spezifikation
P43235 | |
0.1 ng/mL | |
ELISA | |
Colorimetric Microplate Reader | |
CTSK | |
<10% | |
HRP | |
RUO | |
Refer to product documentation for component specific storage temperature. | |
1 hr 20 min |
0.16 to 10 ng/mL | |
HRP | |
Plasma | |
1513 | |
<10% | |
Pre-coated 96 well plate, Standard, Biotinylated Detection Antibody, HRP Conjugate, Standard & Sample Diluent, Biotinylated Detection Antibody Diluent, HRP Conjugate Diluent, Wash Buffer, TMB Substrate, Stop Solution, Plate Sealer | |
96 Tests | |
100 µL | |
Human | |
3 hr 30 min |