missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cathepsin C/DPPI Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Marke: R&D Systems AF1071
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Cathepsin C/DPPI Polyclonal specifically detects Cathepsin C/DPPI in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Spezifikation
| Cathepsin C/DPPI | |
| Polyclonal | |
| Unconjugated | |
| P53634 | |
| CTSC | |
| Mouse myeloma cell line NS0-derived recombinant human pro Cathepsin C/DPPI Asp25-Leu463 Accession # P53634 | |
| 100 μg | |
| Primary | |
| Detects human Cathepsin C/DPPI in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| cathepsin CEC 3.4.14.1, Cathepsin J, CPPIHMS, CTSC, dipeptidyl peptidase 1, Dipeptidyl peptidase I, Dipeptidyl transferase, dipeptidyl-peptidase I, DPP1, DPPI, DPP-I, JP, JPD, PALS, PLS | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1075 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur