missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CASQ1 Full-length ORF (NP_001222.2, 1 a.a. - 390 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00000844-P01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16126017

  • 574.00 CHF / 25 Mikrogramm
Verfügbar ab: 09-01-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

The protein encoded by this gene is a mitochondrial calcium-binding protein located in the luminal space of the terminal cisternae of the sarcoplasmic reticulum. The protein binds and putatively stores calcium ions. The protein is absent in patients with Duchenne and Becker types of muscular dystrophy. [provided by RefSeq]

Sequence: MGPRAVPGLRLALLLLLVLGTPKSGVQGQEGLDFPEYDGVDRVINVNAKNYKNVFKKYEVLALLYHEPPEDDKASQRQFEMEELILELAAQVLEDKGVGFGLVDSEKDAAVAKKLGLTEVDSMYVFKGDEVIEYDGEFSADTIVEFLLDVLEDPVELIEGERELQAFENIEDEIKLIGYFKSKDSEHYKAFEDAAEEFHPYIPFFATFDSKVAKKLTLKLNEIDFYEAFMEEPVTIPDKPNSEEEIVNFVEEHRRSTLRKLKPESMYETWEDDMDGIHIVAFAEEADPDGFEFLETLKAVAQDNTENPDLSIIWIDPDDFPLLVPYWEKTFDIDLSAPQIGVVNVTDADSVWMEMDDEEDLPSAEELEDWLEDVLEGEINTEDDDDDDDD
Spezifikation

Spezifikation

NP_001222.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
70.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CASQ/PDIB1
CASQ1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
844
CASQ1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGPRAVPGLRLALLLLLVLGTPKSGVQGQEGLDFPEYDGVDRVINVNAKNYKNVFKKYEVLALLYHEPPEDDKASQRQFEMEELILELAAQVLEDKGVGFGLVDSEKDAAVAKKLGLTEVDSMYVFKGDEVIEYDGEFSADTIVEFLLDVLEDPVELIEGERELQAFENIEDEIKLIGYFKSKDSEHYKAFEDAAEEFHPYIPFFATFDSKVAKKLTLKLNEIDFYEAFMEEPVTIPDKPNSEEEIVNFVEEHRRSTLRKLKPESMYETWEDDMDGIHIVAFAEEADPDGFEFLETLKAVAQDNTENPDLSIIWIDPDDFPLLVPYWEKTFDIDLSAPQIGVVNVTDADSVWMEMDDEEDLPSAEELEDWLEDVLEGEINTEDDDDDDDD
RUO
CASQ1
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote