Learn More
Abnova™ Human CASP5 Partial ORF (NP_004338, 309 a.a. - 418 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
This gene encodes a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. Overexpression of the active form of this enzyme induces apoptosis in fibroblasts. Max, a central component of the Myc/Max/Mad transcription regulation network important for cell growth, differentiation, and apoptosis, is cleaved by this protein; this process requires Fas-mediated dephosphorylation of Max. The expression of this gene is regulated by interferon-gamma and lipopolysaccharide. Several alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_004338 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 838 |
Molekulargewicht | 37.84kDa |
Name | CASP5 (Human) Recombinant Protein (Q01) |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 µg |
Immunogen | VRDSPASLAVISSQSSENLEADSVCKIHEEKDFIAFCSSTPHNVSWRDRTRGSIFITELITCFQKYSCCCHLMEIFRKVQKSFEVPQAKAQMPTIERATLTRDFYLFPGN |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.