Learn More
Abnova™ Human CARD14 Full-length ORF (NP_438170.1, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal
Description
The protein encoded by this gene belongs to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. This protein is also a member of the CARD protein family, which is defined by carrying a characteristic caspase-associated recruitment domain (CARD). This protein shares a similar domain structure with CARD11 protein. The CARD domains of both proteins have been shown to specifically interact with BCL10, a protein known to function as a positive regulator of cell apoptosis and NF-kappaB activation. When expressed in cells, this protein activated NF-kappaB and induced the phosphorylation of BCL10. Two alternatively spliced variants of this gene encoding distinct isoforms have been reported. [provided by RefSeq]
Specifications
Specifications
Zugriffsnummer | NP_438170.1 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 79092 |
Molekulargewicht | 74.9kDa |
Name | CARD14 (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 μg |
Immunogen | MVSSCELELQEQSLRTASDQESGDEELNRLKEENEKLRSLTFSLAEKDILEQSLDEARGSRQELVERIHSLRERAVAAERQREQYWEEKEQTLLQFQKSKMACQLYREKVNALQAQVCELQKERDQAYSARDSAQREISQSLVEKDSLRRQVFELTDQVCELRTQLRQLQAEPPGVLKQEARTREPCPREKQRLVRMHAICPRDDSDCSLVSSTESQLLSDLSATSSRELVDSFRSSSPAPPSQQSLYKRVAEDFGEEPWSFSSCLEIPEGDPGALPGAKAGDPHLDYELLDTADLPQLESSLQPVSPGRLDVSESGVLMRRRPARRILSQVTMLAFQGDALLEQISVIGGNLTGIFIHRVTPGSAADQMALRPGTQIVMVSRARPLLSPGLLMGTVAAGGVTQADFTSPRRCRSTLGWASALSWADVKRSAHL |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.