Learn More
Invitrogen™ Human Carbonic Anhydrase I (aa 12-150) Control Fragment Recombinant Protein
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (79%), Rat (79%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82254 (PA5-82254. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Spezifikation
Spezifikation
| Zugriffsnummer | P00915 |
| Konzentration | ≥5.0 mg/mL |
| Zur Verwendung mit (Anwendung) | Blocking Assay, Control |
| Zusammensetzung | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-ID (Entrez) | 759 |
| Name | Human Carbonic Anhydrase I (aa 12-150) Control Fragment |
| Menge | 100 μl |
| Kennzeichnung | RUO |
| Gen-Alias | AW555628; CA I; CA1; ca1 {ECO:0000250; CAB; CA-I; CA-I {ECO:0000250; Car1; Car-1; Carbonate dehydratase I; carbonate dehydratase I {ECO:0000250; carbonic anhydrase 1; carbonic anhydrase 1 {ECO:0000250; Carbonic anhydrase B; carbonic anhydrase I; carbonic anhydrase I {ECO:0000250; epididymis secretory protein Li 11; HEL-S-11; UniProtKB:P00915} |
| Gebräuchliche Bezeichnung | Carbonic Anhydrase I |
| Mehr anzeigen |