Learn More
Abnova™ Human C1orf48 Partial ORF (NP_056286, 182 a.a. - 279 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_056286 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 25936 |
Molekulargewicht | 36.52kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16125216
|
Abnova™
H00025936-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 31-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16135216
|
Abnova™
H00025936-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 31-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a protein with two coiled-coil domains that localizes to kinetochores, which are chromosome-associated structures that attach to microtubules and mediate chromosome movements during cell division. The encoded protein is part of a conserved protein complex that includes two chromodomain-containing proteins and a component of the outer plate of the kinetochore. This protein complex is proposed to bridge centromeric heterochromatin with the outer kinetochore structure. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: KEISEAMKSLPALIEQGEGFSQVLRMQPVIHLQRIHQEVFSSCHRKPDAKPENFITQIETTPTETASRKTSDVVLKRKQTKDCPQRKWYPLRPKKINLSpezifikation
NP_056286 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C1orf48/DC8/DKFZp566O1646/MIS14 | |
NSL1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
25936 | |
C1orf48 (Human) Recombinant Protein (Q01) | |
KEISEAMKSLPALIEQGEGFSQVLRMQPVIHLQRIHQEVFSSCHRKPDAKPENFITQIETTPTETASRKTSDVVLKRKQTKDCPQRKWYPLRPKKINL | |
RUO | |
NSL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |