missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C1orf181 Full-length ORF (AAH26236.1, 1 a.a. - 470 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00054680-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MEFAAENEGKSGGGLHSVAEGVRLSPEPGREGVRDLAGAEEFGGGEEGTGLTGIKEIGDGEEGSGQRPEEIPMDLTVVKQEIIDWPGTEGRLAGQWVEQEVEDRPEVKDENAGVLEVKQETDSSLVVKEAKVGEPEVKEEKVKEEVMDWSEVKEEKDNLEIKQEEKFVGQCIKEELMHGECVKEEKDFLKKEIVDDTKVKEEPPINHPVGCKRKLAMSRCETCGTEEAKYRCPRCMRYSCSLPCVKKHKAELTCNGVRDKTAYISIQQFTEMNLLSDYRFLEDVARTADHISRDAFLKRPISNKYMYFMKNRARRQGINLKLLPNGFTKRKENSTFFDKKKQQFCWHVKLQFPQSQAEYIEKRVPDDKTINEILKPYIDPEKSDPVIRQRLKAYIRSQTGVQILMKIEYMQQNLVRYYELDPYKSLLDNLRNKVIIEYPTLHVVLKGSNNDMKVLHQVKSESTKNVGNENSpezifikation
AAH26236.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
80.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BCD1/C1orf181/FLJ20729/FLJ20760/MGC131963/NY-BR-75 | |
ZNHIT6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54680 | |
C1orf181 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEFAAENEGKSGGGLHSVAEGVRLSPEPGREGVRDLAGAEEFGGGEEGTGLTGIKEIGDGEEGSGQRPEEIPMDLTVVKQEIIDWPGTEGRLAGQWVEQEVEDRPEVKDENAGVLEVKQETDSSLVVKEAKVGEPEVKEEKVKEEVMDWSEVKEEKDNLEIKQEEKFVGQCIKEELMHGECVKEEKDFLKKEIVDDTKVKEEPPINHPVGCKRKLAMSRCETCGTEEAKYRCPRCMRYSCSLPCVKKHKAELTCNGVRDKTAYISIQQFTEMNLLSDYRFLEDVARTADHISRDAFLKRPISNKYMYFMKNRARRQGINLKLLPNGFTKRKENSTFFDKKKQQFCWHVKLQFPQSQAEYIEKRVPDDKTINEILKPYIDPEKSDPVIRQRLKAYIRSQTGVQILMKIEYMQQNLVRYYELDPYKSLLDNLRNKVIIEYPTLHVVLKGSNNDMKVLHQVKSESTKNVGNEN | |
RUO | |
ZNHIT6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |