missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human C13orf7 Partial ORF (NP_078822, 1 a.a. - 95 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00079596-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: MLSHTVRKHLRKTRLELLHKEYEDEIDCLQKEVEELKSKNLSLESQIKTILDPLTLVQGNQNEDKHLVTDNPSKINPETVAEWKKKLRTANEIYESpezifikation
NP_078822 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.19kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLSHTVRKHLRKTRLELLHKEYEDEIDCLQKEVEELKSKNLSLESQIKTILDPLTLVQGNQNEDKHLVTDNPSKINPETVAEWKKKLRTANEIYE | |
RUO | |
RNF219 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
79596 | |
C13orf7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C13orf7/DKFZp686A01276/DKFZp686N15250/DKFZp686O03173/FLJ13449/FLJ25774 | |
RNF219 | |
Recombinant | |
wheat germ expression system |