missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human BTN1A1 / Butyrophilin Protein, Fc Tag

362.66 CHF - 2501.16 CHF

Spezifikation

Zugriffsnummer AAH96312
Molekulargewicht 50.6 kDa
Quelle Human BTN1A1, Fc Tag (BT1-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 242 (Accession # AAH96312).
Immunogen Ala 27 - Arg 242
Lagerungsbedingungen -20°C
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15836057
Unterlagen ansehen Aktionsdetails
ACROBiosystems
BT1-H5253-1MG
1 mg
2501.16 CHF
1 Milligramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15826057
Unterlagen ansehen Aktionsdetails
ACROBiosystems
BT1-H5253-100UG
100 μg
362.66 CHF
100 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Butyrophilin subfamily 1 member A1 (BTN1A1) is also known as BTN, which is a member of the immunoglobulin superfamily and the major protein associated with fat droplets in the milk. BTN1A1 may have a cell surface receptor function. The human butyrophilin gene is localized in the major histocompatibility complex (MHC) class I region of 6p and may have arisen relatively recently in evolution by the shuffling of exons between 2 ancestral gene families. Furthermore, BTN1A1 regulates the amount of lipids and size of droplets expressed in milk and inhibits the proliferation of CD4 and CD8 T-cells activated by anti-CD3 antibodies, T-cell metabolism and IL2 and IFNG secretion.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50.6 kDa. As a result of glycosylation, the protein migrates as 55-66 kDa under reducing (R) condition, and 116-130 kDa under non-reducing (NR) condition (SDS-P
Spezifikation

Spezifikation

AAH96312
Human BTN1A1, Fc Tag (BT1-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Arg 242 (Accession # AAH96312).
-20°C
BTN1A1
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Immune Checkpoint Proteins
50.6 kDa
Ala 27 - Arg 242
BTN1A1,Butyrophilin,BTN
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications