Learn More
ACROBiosystems Human BTLA (31-134) Protein, Fc Tag
Marke:  ACROBiosystems BTA-H5255-100UG
Additional Details : Gewicht : 0.00001kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15826067
  • 441.35 CHF / 100 Mikrogramm
Verfügbar ab: 07-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 38.7 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation
Spezifikation
NP_001078826.1
100 μg
Lys 31 - Thr 134
BTLA,CD272
Unconjugated
Fc Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>92% as determined by SDS-PAGE
38.7 kDa
Human BTLA, Fc Tag (BTA-H5255) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Thr 134 (Accession # NP_001078826.1).
-20°C
BTLA
Human
Tris with Glycine, Arginine and NaCl, pH7.5
HEK293
Immune Checkpoint Proteins