Learn More
ACROBiosystems Human BTLA (31-134) Protein, Fc Tag
Marke: ACROBiosystems BTA-H5255-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
B- and T-lymphocyte attenuator (BTLA) is also known as B- and T-lymphocyte-associated protein, CD antigen CD272. BTLA contains one Ig-like V-type (immunoglobulin-like) domain. As a lymphocyte inhibitory receptor, BTLA / CD272 inhibits lymphocytes during immune response. BTLA / CD272 can interact with tyrosine phosphatases PTPN6/SHP-1 and PTPN11/SHP-2, and interact with TNFRSF14/HVEM.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 38.7 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001078826.1 | |
1 mg | |
Lys 31 - Thr 134 | |
BTLA,CD272 | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
38.7 kDa | |
Human BTLA, Fc Tag (BTA-H5255) is expressed from human 293 cells (HEK293). It contains AA Lys 31 - Thr 134 (Accession # NP_001078826.1). | |
-20°C | |
BTLA | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |