missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human BSG Partial ORF (NP_001719, 206 a.a. - 305 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
361.00 CHF - 547.00 CHF
Spezifikation
Zugriffsnummer | NP_001719 |
---|---|
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 682 |
Molekulargewicht | 36.74kDa |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16132351
|
Abnova™
H00000682-Q01.25UG |
25 ug |
547.00 CHF
25 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16122351
|
Abnova™
H00000682-Q01.10UG |
10 ug |
361.00 CHF
10 Mikrogramm |
Verfügbar ab: 05-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
The protein encoded by this gene is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. The encoded protein is also a member of the immunoglobulin superfamily. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: LPEPMGTANIQLHGPPRVKAVKSSEHINEGETAMLVCKSESVPPVTDWAWYKITDSEDKALMNGSESRFFVSSSQGRSELHIENLNMEADPGQYRCNGTSSpezifikation
NP_001719 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
5F7/CD147/EMMPRIN/M6/OK/TCSF | |
BSG | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
682 | |
BSG (Human) Recombinant Protein (Q01) | |
LPEPMGTANIQLHGPPRVKAVKSSEHINEGETAMLVCKSESVPPVTDWAWYKITDSEDKALMNGSESRFFVSSSQGRSELHIENLNMEADPGQYRCNGTS | |
RUO | |
BSG | |
Wheat Germ (in vitro) | |
GST | |
Liquid |