missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human BRP44L (aa 85-107) Control Fragment Recombinant Protein

Artikelnummer. 30200428
Change view
Click to view available options
Menge:
100 μl
Packungsgröße:
100 Mikroliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30200428 100 μl 100 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30200428 Lieferant Invitrogen™ Lieferanten-Nr. RP106663

om dit product te kopen Registreer vandaag om een webaccount aan te maken

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (74%), Rat (74%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84267 (PA5-84267. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

BRP44L (brain protein 44-like protein), also known as HSPC040 or CGI-129, is a 109 amino acid mitochondrial protein belonging to the UPF0041 family. The gene that encodes BRP44L maps to human chromosome 6. Making up nearly 6% of the human genome, chromosome 6 contains around 1,200 genes within 170 million base pairs of sequence. Deletion of a portion of the q arm of chromosome 6 is associated with early onset intestinal cancer suggesting the presence of a cancer susceptibility locus. Porphyria cutanea tarda is associated with chromosome 6 through the HFE gene which, when mutated, predisposes an individual to developing this porphyria. Notably, the PARK2 gene, which is associated with Parkinson's disease, and the genes encoding the major histocompatiblity complex proteins, which are key molecular components of the immune system and determine predisposition to rheumatic diseases, are also located on chromosome 6. Stickler syndrome, 21-hydroxylase deficiency and maple syrup urine disease are also associated with genes on chromosome 6. A bipolar disorder susceptibility locus has been identified on the q arm of chromosome 6.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer Q9Y5U8
Konzentration ≥5.0 mg/mL
Zur Verwendung mit (Anwendung) Blocking Assay, Control
Zusammensetzung 1 M urea, PBS with no preservative; pH 7.4
Gen-ID (Entrez) 51660
Name Human BRP44L (aa 85-107) Control Fragment
Menge 100 μl
Kennzeichnung RUO
Gen-Alias 0610006G08Rik; 3830411I18Rik; apoptosis-regulating basic protein; Arbp; Brain protein 44-like protein; Brp44l; CGI-129; dJ68L15.3; HSPC040; HSPC040 protein; mitochondrial pyruvate carrier 1; Mpc1; MPYCD; PNAS-115
Gebräuchliche Bezeichnung BRP44L
Gensymbol MPC1
Konjugat Unconjugated
Spezies Human
Rekombinant Recombinant
Proteinmarkierung His-ABP-tag
Sequenz ATNEVAQLIQGGRLIKHEMTKTA
Inhalt und Lagerung -20°C, Avoid Freeze/Thaw Cycles
Expressionssystem E. coli
Form Liquid
Reinheits- oder Qualitätsgrad >80% by SDS-PAGE and Coomassie blue staining
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.