Learn More
Abnova™ Human BLID Full-length ORF (NP_001001786.1, 1 a.a. - 108 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00414899-P01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a BH3-like motif containing protein involved in cell death. The encoded protein may induce apoptosis in a caspase-dependent manner. The protein is localized in both the cytoplasm and the mitochondrion. [SH
Sequence: MVTLLPIEGQEVHFFEILESECVLYTGWIERASGSSIYPEAKARLPLEALLGSNKEPMLPKETVLSLKRYNLGSSAMKRNVPGHVLQRPSYLTRIQVTLLCNSSAEALSpezifikation
NP_001001786.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
38.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
BLID | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
414899 | |
BLID (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BRCC2/BRCC@/MGC163233/MGC163235 | |
BLID | |
Recombinant | |
wheat germ expression system |