Learn More
ACROBiosystems Human Bcl-x / BCL2L1 Protein, GST Tag
Marke: ACROBiosystems BC1-H513A-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Bcl-2-like protein 1 (BCL2L1), is also known as Apoptosis regulator Bcl-X (Bcl-XL). BCL2L1 / Bcl-XL is a homodimer protein and belongs to the Bcl-2 family. BCL2L1 is expressed at high levels in cells that undergo a high rate of turnover, such as developing lymphocytes. BCL2L1 is a potent inhibitor of cell death. BCL2L1 Inhibits activation of caspases. BCL2L1 appears to regulate cell death by blocking the voltage-dependent anion channel (VDAC) by binding to it and preventing the release of the caspase activator, CYC1, from the mitochondrial membrane. BCL2L1 / Bcl-XL also acts as a regulator of G2 checkpoint and progression to cytokinesis during mitosis.
This protein carries a GST tag at the N-terminus. The protein has a calculated MW of 50.6 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE).Spezifikation
AAH19307 | |
1 mg | |
Ser 2 - Arg 212 | |
BCL2L1,Bcl-XL,Bcl2-L-1,BCL-XL/S,BCL2L,BCLX,BCLXL,BCLXS,Bcl-X,bcl-xS | |
Unconjugated | |
GST Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>92% as determined by SDS-PAGE |
50.6 kDa | |
Human Bcl-x, GST Tag (BC1-H513a) is expressed from E.coli cells. It contains AA Ser 2 - Arg 212 (Accession # AAH19307). | |
-20°C | |
Bcl-x | |
Human | |
20 mM Tris, 100 mM NaCl, pH7.5 with 10 mM GSH | |
E.coli | |
Bio-Markers and CD Antigens |