missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Human B7-H5 / Gi24 / VISTA Protein, His Tag

MALS verified

Marke:  ACROBiosystems B75-H52H0-100UG

Additional Details : Gewicht : 0.00001kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 15825977

  • 322.42 CHF / 100 Mikrogramm


Verfügbar ab
28-05-2021
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Platelet receptor Gi24, also known as B7-H5 and stress-induced secreted protein-1 (Sisp-1), is a protein that in humans is encoded by the C10orf54 gene, which contains 1 Ig-like (immunoglobulin-like) domain. As for C10orf54 gene, C10orf54 appears to positively interact with BMP-4, potentiating BMP signaling and the transition from an undifferentiated to a differentiated state on ESCs. Human C10orf54 undergoes proteolytic cleavage by MT1-MMP, generating a soluble 30 kDa extracellular fragment plus a 25-30 kDa membrane-bound fragment.

This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.0 kDa. The protein migrates as 33-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spezifikation

Spezifikation

AAH20568
HEK293
Human
20.0 kDa
100 µg
Phe 33 - Ala 194
B7-H5,SISP1,Gi24,VISTA
Unconjugated
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>90% as determined by SDS-PAGE
Human B7-H5, His Tag (B75-H52H0) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568).
-20°C
B7-H5
His Tag
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation