missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human B7-H2 Fc Recombinant Protein, PeproTech®

Artikelnummer. 17822473
Change view
Click to view available options
Menge:
1 mg
100 μg
20 μg
250 μg
500 μg
Packungsgröße:
1 Milligramm
100 Mikrogramm
20 Mikrogramm
250 Mikrogramm
500 Mikrogramm
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17822473 100 μg 100 Mikrogramm
17802293 1 mg 1 Milligramm
17872063 20 μg 20 Mikrogramm
17862993 250 μg 250 Mikrogramm
17852873 500 μg 500 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17822473 Lieferant Gibco™ Lieferanten-Nr. 31037100UG

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

PeproTech's CHO cell-derived Recombinant Human B7-H2 Fc is a glycosylated, disulfide-linked homodimer of 942 amino acid residues whose monomer consists of the 238-amino-acid length extracellular portion of B7-H2 fused to the 231-amino-acid length Fc portion of human IgG1 by two glycines. The calculated molecular weight of Recombinant Human B7-H2 Fc dimer is 104.9 kDa; however, due to glycosylation, it migrates at an apparent molecular weight of approximately 90-95 kDa by SDS-PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Inducible co-stimulator ligand (ICOSL) is a specific ligand on antigen-presenting cells and cells of the peripheral tissue that binds to the inducible co-stimulator receptor (ICOS). ICOS belongs to the CD28/CD152 receptor family that regulates T-cell activation and function. ICOSL is expressed on monocytes, dendritic cells and B cells and can be induced by inflammatory stimuli in peripheral tissue. Binding to ICOSL delivers a co-stimulatory signal for T cell proliferation and cytokine secretion.
TRUSTED_SUSTAINABILITY

Spezifikation

Zugriffsnummer O75144
Zur Verwendung mit (Anwendung) Functional Assay
Zusammensetzung protein with no preservative
Gen-ID (Entrez) 23308
Molekulargewicht 104.9 kDa
Name Human B7-H2 Fc
Menge 100 μg
Quelle CHO cells
Kennzeichnung RUO
Endotoxin-Konzentration <1 EU/ μg
Gen-Alias AU044799; B7 homolog 2; B7 homologue 2; B7h; B7H2; B7-H2; B7-like protein Gl50; B7-related protein 1; B7RP1; B7RP-1; BG071784; CD275; GI50; GL50; GL50-B; ICOS ligand; ICOSL; ICOS-L; ICOSLG; inducible T-cell costimulator ligand; inducible T-cell co-stimulator ligand; KIAA0653; LICOS; Ly115l; mKIAA0653; RGD1562791; transmembrane protein B7-H2 ICOS ligand
Gebräuchliche Bezeichnung CD275 (B7-H2)
Gensymbol ICOSLG
Biologische Aktivität Determined by its ability to induce adhesion in Jurkat cells activated by immobilized anti-CD3. The ED50 for this effect is 0.3-0.6 ug/ml
Konjugat Unconjugated
Rekombinant Recombinant
Proteinmarkierung Fc-tag
Sequenz DTQEKEVRAM VGSDVELSCA CPEGSRFDLN DVYVYWQTSE SKTVVTYHIP QNSSLENVDS RYRNRALMSP AGMLRGDFSL RLFNVTPQDE QKFHCLVLSQ SLGFQEVLSV EVTLHVAANF SVPVVSAPHS PSQDELTFTC TSINGYPRPN VYWINKTDNS LLDQALQNDT VFLNMRGLYD VVSVLRIART PSVNIGCCIE NVLLQQNLTV GSQTGNDIGE RDKITENPVS TGEKNAATGG PKSCDKTHTC PPCPAPELLG GPSVFLFPPK PKDTLMISRT PEVTCVVVDV SHEDPEVKFN WYVDGVEVHN AKTKPREEQY NSTYRVVSVL TVLHQDWLNG KEYKCKVSNK ALPAPIEKTI SKAKGQPREP QVYTLPPSRD ELTKNQVSLT CLVKGFYPSD IAVEWESNGQ PENNYKTTPP VLDSDGSFFL YSKLTVDKSR WQQGNVFSCS VMHEALHNHY TQKSLSLSPG K
Inhalt und Lagerung -20°C
Expressionssystem CHO cells
Form Lyophilized
Reinheits- oder Qualitätsgrad ≥ 95% by SDS-PAGE gel and HPLC analyses.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.