Learn More
ACROBiosystems Human B7-2 / CD86 Protein, His Tag
353.08 CHF - 2383.29 CHF
Spezifikation
Zugriffsnummer | AAH40261 |
---|---|
Molekulargewicht | 26.2 kDa |
Quelle | Human B7-2, His Tag (CD6-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accession # AAH40261). |
Immunogen | Leu 26 - Pro 247 |
Lagerungsbedingungen | -20°C |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15826227
|
ACROBiosystems
CD6-H5223-100UG |
100 μg |
353.08 CHF
100 Mikrogramm |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
15836227
|
ACROBiosystems
CD6-H5223-1MG |
1 mg |
2383.29 CHF
1 Milligramm |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Cluster of Differentiation 86 (CD86) is also known as B-lymphocyte activation antigen B7-2, is a type I membrane protein that is a member of the immunoglobulin superfamily, and is constitutively expressed on interdigitating dendritic cells, Langerhans cells, peripheral blood dendritic cells, memory B cells, and germinal center B cells. Additionally, B72 is expressed at low levels on monocytes and can be upregulated through interferon γ. CD86 is the ligand for two different proteins on the T cell surface: CD28 (for autoregulation and intercellular association) and CTLA-4 (for attenuation of regulation and cellular disassociation). CD86 works in tandem with CD80 to prime T cells. Recent study has revealed that B7-2 promotes the generation of a mature APC repertoire and promotes APC function and survival. Furthermore, the B7 proteins are also involved in innate immune responses by activating NF-κB-signaling pathway in macrophages. CD86 thus is regarded as a promising candidate for immune therapy. CD86+ macrophages in Hodgkin lymphoma patients are an independent marker for potential nonresponse to firstline-therapy.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.2 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
AAH40261 | |
Human B7-2, His Tag (CD6-H5223) is expressed from human 293 cells (HEK293). It contains AA Leu 26 - Pro 247 (Accession # AAH40261). | |
-20°C | |
B7-2 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
26.2 kDa | |
Leu 26 - Pro 247 | |
CD86,B7-2,B70,CD28LG2,LAB72,MGC34413 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |