Learn More
Abnova™ Human ATP6V1B1 Partial ORF (NP_001683.2, 1 a.a. - 75 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c″, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is one of two V1 domain B subunit isoforms and is found in the kidney. Mutations in this gene cause distal renal tubular acidosis associated with sensorineural deafness. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_001683.2 |
Zur Verwendung mit (Anwendung) | Antibody Production, ELISA, Protein Array, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 525 |
Molekulargewicht | 33.99kDa |
Name | ATP6V1B1 (Human) Recombinant Protein (Q01) |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 10 µg |
Immunogen | MAMEIDSRPGGLPGSSCNLGAAREHMQAVTRNYITHPRVTYRTVCSVNGPLVVLDRVKFAQYAEIVHFTLPDGTQ |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.