missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ATP6V1B1 Partial ORF (NP_001683.2, 1 a.a. - 75 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

361.00 CHF - 547.00 CHF

Spezifikation

Zugriffsnummer NP_001683.2
Zur Verwendung mit (Anwendung) Antibody Production, ELISA, Protein Array, Western Blot
Zusammensetzung 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gen-ID (Entrez) 525
Molekulargewicht 33.99kDa
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
16135074
Unterlagen ansehen Aktionsdetails
Abnova™
H00000525-Q01.10UG
10 ug
361.00 CHF
10 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
16145074
Unterlagen ansehen Aktionsdetails
Abnova™
H00000525-Q01.25UG
25 ug
547.00 CHF
25 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 10-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c', c″, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is one of two V1 domain B subunit isoforms and is found in the kidney. Mutations in this gene cause distal renal tubular acidosis associated with sensorineural deafness. [provided by RefSeq]

Sequence: MAMEIDSRPGGLPGSSCNLGAAREHMQAVTRNYITHPRVTYRTVCSVNGPLVVLDRVKFAQYAEIVHFTLPDGTQ
Spezifikation

Spezifikation

NP_001683.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
33.99kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATP6B1/MGC32642/RTA1B/VATB/VMA2/VPP3
ATP6V1B1
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
525
ATP6V1B1 (Human) Recombinant Protein (Q01)
MAMEIDSRPGGLPGSSCNLGAAREHMQAVTRNYITHPRVTYRTVCSVNGPLVVLDRVKFAQYAEIVHFTLPDGTQ
RUO
ATP6V1B1
Wheat Germ (in vitro)
GST
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications