missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Arylsulfatase B/ARSB Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Marke: R&D Systems MAB4415-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Arylsulfatase B/ARSB Monoclonal specifically detects Arylsulfatase B/ARSB in Human samples. It is validated for Western Blot, Immunocytochemistry.
Spezifikation
| Arylsulfatase B/ARSB | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P15848 | |
| ARSB | |
| Chinese hamster ovary cell line CHO-derived recombinant human Arylsulfatase B/ARSB Ser37-Met533 Accession # P15848 | |
| 25 μg | |
| Primary | |
| Detects human Arylsulfatase B/ARSB in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ARSA, rhARSJ, or recombinant mouse ARSG is observed. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 708401 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARSB, arylsulfatase B, ASB, EC 3.1.6.12, G4S, MPS6, N-acetylgalactosamine-4-sulfatase | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 411 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur