missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ARNTL2 Partial ORF (NP_064568, 285 a.a. - 379 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00056938-Q01.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: RIKSCKISVKEEHGCLPNSKKKEHRKFYTIHCTGYLRSWPPNIVGMEEERNSKKDNSNFTCLVAIGRLQPYIVPQNSGEINVKPTEFITRFAVNGSpezifikation
NP_064568 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.19kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RIKSCKISVKEEHGCLPNSKKKEHRKFYTIHCTGYLRSWPPNIVGMEEERNSKKDNSNFTCLVAIGRLQPYIVPQNSGEINVKPTEFITRFAVNG | |
RUO | |
ARNTL2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
56938 | |
ARNTL2 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BMAL2/CLIF/MGC149671/MGC149672/MOP9/PASD9/bHLHe6 | |
ARNTL2 | |
Recombinant | |
wheat germ expression system |