missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ARNT2 Partial ORF (NP_055677.3, 464 a.a. - 563 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marke:  Abnova™ H00009915-Q01.25ug

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16151446

  • 547.00 CHF / 25 Mikrogramm
Verfügbar ab: 13-06-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a member of the basic-helix-loop-helix-Per-Arnt-Sim (bHLH-PAS) superfamily of transcription factors. The encoded protein acts as a partner for several sensor proteins of the bHLH-PAS family, forming heterodimers with the sensor proteins that bind regulatory DNA sequences in genes responsive to developmental and environmental stimuli. Under hypoxic conditions, the encoded protein complexes with hypoxia-inducible factor 1alpha in the nucleus and this complex binds to hypoxia-responsive elements in enhancers and promoters of oxygen-responsive genes. A highly similar protein in mouse forms functional complexes with both aryl hydrocarbon receptors and Single-minded proteins, suggesting addition roles for the encoded protein in the metabolism of xenobiotic compounds and the regulation of neurogenesis, respectively. [provided by RefSeq]

Sequence: VPVPNLPAGVHEAGKSVEKADAIFSQERDPRFAEMFAGISASEKKMMSSASAAGTQQIYSQGSPFPSGHSGKAFSSSVVHVPGVNDIQSSSSTGQNMSQI
Spezifikation

Spezifikation

NP_055677.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VPVPNLPAGVHEAGKSVEKADAIFSQERDPRFAEMFAGISASEKKMMSSASAAGTQQIYSQGSPFPSGHSGKAFSSSVVHVPGVNDIQSSSSTGQNMSQI
RUO
ARNT2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9915
ARNT2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0307/bHLHe1
ARNT2
Recombinant
wheat germ expression system
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote