Learn More
ACROBiosystems Human APCS / SAP / PTX2 Protein, His Tag
Marke: ACROBiosystems APS-H5229-1MG
Additional Details : Gewicht : 0.00001kg
Beschreibung
Serum amyloid P-component (SAP) is also known as Pentraxin 2 (PTX2), APCS, which belongs to the pentaxin family. SAP contains one pentaxin domain. PTX2 is a secreted homopentamer protein which is found in serum and urine. SAP is a precursor of amyloid component P which is found in basement membrane and associated with amyloid deposits. SAP can interact with DNA and histones and may scavenge nuclear material released from damaged circulating cells. SAP may also function as a calcium-dependent lectin. SAP has certain sequence homology with C-reactive protein (CRP).
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.1 kDa. The protein migrates as 26 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
NP_001630.1 | |
1 mg | |
His 20 - Val 223 | |
PTX2,APCS,SAMP,SAP,Pentraxin 2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
24.1 kDa | |
Human APCS, His Tag (APS-H5229) is expressed from human 293 cells (HEK293). It contains AA His 20 - Val 223 (Accession # P02743-1). | |
-20°C | |
APCS | |
Human | |
50 mM Tris, 150 mM NaCl, pH 7.5 | |
HEK293 | |
Other Recombinant Proteins |