Learn More
R&D Systems™ Human Angiopoietin-1 (NP_001137.2) VersaClone cDNA
Spezifikation
Spezifikation
| Gen | Angiopoietin-1 |
| Zugriffsnummer | NP_001137.2 |
| Gen-ID (Entrez) | 284 |
| Gen-Alias | AGP1, AGPT, Ang1, ANG-1, angiopoietin 1, angiopoietin-1, KIAA0003 |
| Inhalt und Lagerung | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Produktlinie | R&D Systems |
| Menge | 10 μg |