Learn More
Abnova™ Human ALPP Full-length ORF (NP_001623.3, 1 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal
Beschreibung
There are at least four distinct but related alkaline phosphatases: intestinal, placental, placental-like, and liver/bone/kidney (tissue non-specific). The first three are located together on chromosome 2 while the tissue non-specific form is located on chromosome 1. The product of this gene is a membrane bound glycosylated enzyme, also referred to as the heat stable form, that is expressed primarily in the placenta although it is closely related to the intestinal form of the enzyme as well as to the placental-like form. The coding sequence for this form of alkaline phosphatase is unique in that the 3' untranslated region contains multiple copies of an Alu family repeat. In addition, this gene is polymorphic and three common alleles (type 1, type 2 and type 3) for this form of alkaline phosphatase have been well characterized. [provided by RefSeq]
Spezifikation
Spezifikation
Zugriffsnummer | NP_001623.3 |
Zur Verwendung mit (Anwendung) | Antibody Production, Protein Array, ELISA, Western Blot |
Zusammensetzung | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gen-ID (Entrez) | 250 |
Molekulargewicht | 84.4kDa |
Name | ALPP (Human) Recombinant Protein (P01) |
Reinigungsverfahren | Glutathione Sepharose 4 Fast Flow |
Qualitätskontrollen | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Menge | 25 μg |
Lagerungsbedingungen | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Mehr anzeigen |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.