Learn More
Abnova™ Human ALOX12B Full-length ORF (NP_001130.1, 1 a.a. - 701 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00000242-P01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes an enzyme involved in the converstion of arachidonic acid to 12R-hydroxyeicosatetraenoic acid. Mutations in this gene are associated with nonbullous congenital ichthyosiform erythroderma. [provided by RefSeq]
Sequence: MATYKVRVATGTDLLSGTRDSISLTIVGTQGESHKQLLNHFGRDFATGAVGQYTVQCPQDLGELIIIRLHKERYAFFPKDPWYCNYVQICAPNGRIYHFPAYQWMDGYETLALREATGKTTADDSLPVLLEHRKEEIRAKQDFYHWRVFLPGLPSYVHIPSYRPPVRRHRNPNRPEWNGYIPGFPILINFKATKFLNLNLRYSFLKTASFFVRLGPMALAFKVRGLLDCKHSWKRLKDIRKIFPGKKSVVSEYVAEHWAEDTFFGYQYLNGVNPGLIRRCTRIPDKFPVTDDMVAPFLGEGTCLQAELEKGNIYLADYRIMEGIPTVELSGRKQHHCAPLCLLHFGPEGKMMPIAIQLSQTPGPDCPIFLPSDSEWDWLLAKTWVRYAEFYSHEAIAHLLETHLIAEAFCLALLRNLPMCHPLYKLLIPHTRYTVQINSIGRAVLLNEGGLSAKGMSLGVEGFAGVMVRALSELTYDSLYLPNDFVERGVQDLPGYYYRDDSLAVWNALEKYVTEIITYYYPSDAAVEGDPELQSWVQEIFKECLLGRESSGFPRCLRTVPELIRYVTIVIYTCSAKHAAVNTGQMEFTAWMPNFPASMRNPPIQTKGLTTLETFMDTLPDVKTTCITLLVLWTLSREPDDRRPLGHFPDIHFVEEAPRRSIEAFRQRLNQISHDIRQRNKCLPIPYYYLDPVLIENSISISpezifikation
NP_001130.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
106.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
ALOX12B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
242 | |
ALOX12B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
12R-LOX | |
ALOX12B | |
Recombinant | |
wheat germ expression system |