missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Aldehyde Dehydrogenase 1-A1/ ALDH1A1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Marke: R&D Systems MAB5869
Additional Details : Gewicht : 0.00970kg
Beschreibung
Aldehyde Dehydrogenase 1-A1/ALDH1A1 Monoclonal specifically detects Aldehyde Dehydrogenase 1-A1/ALDH1A1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry, CyTOF-reported.Spezifikation
Aldehyde Dehydrogenase 1-A1/ALDH1A1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P00352 | |
ALDH1A1 | |
E. coli-derived recombinant human Aldehyde Dehydrogenase 1-A1/ALDH1-A1 Ser2-Ser501 Accession # P00352 | |
100 μg | |
Primary | |
Detects human Aldehyde Dehydrogenase 1-A1/ALDH1-A1 in direct ELISAs and Western blots. In direct ELISAs, approximatly 50% cross-reactivity with recombinant human Aldehyde Dehydrogenase 1-A2 is observed. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
703410 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
acetaldehyde dehydrogenase 1, ALDC, aldehyde dehydrogenase 1 family, member A1, aldehyde dehydrogenase 1, soluble, Aldehyde Dehydrogenase 1A1, Aldehyde dehydrogenase family 1 member A1, Aldehyde dehydrogenase, cytosolic, aldehyde dehydrogenase, liver cytosolic, ALDH class 1, ALDH1, ALDH11, ALDH1A1, ALDH-E1, ALHDII, EC 1.2.1, EC 1.2.1.36, MGC2318, PUMB1, RALDH 1, RalDH1, retinal dehydrogenase 1, retinaldehyde dehydrogenase 1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
216 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |