missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AK7 Partial ORF (NP_689540, 31 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00122481-Q01.25ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Sequence: GNIGKFLSNCVVGASLEEITEEEEEEDENKSAMLEASSTKVKEGTFQIVGTLSKPDSPRPDFAVETYSAISQEDLLMRLLECDVIIYNITESSQQMEEAISpezifikation
NP_689540 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
GNIGKFLSNCVVGASLEEITEEEEEEDENKSAMLEASSTKVKEGTFQIVGTLSKPDSPRPDFAVETYSAISQEDLLMRLLECDVIIYNITESSQQMEEAI | |
RUO | |
AK7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
122481 | |
AK7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ32864 | |
AK7 | |
Recombinant | |
wheat germ expression system |