Learn More
Abnova™ Human AK3L1 Full-length ORF (NP_001002921.1, 1 a.a. - 223 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00000205-P02.25ug
Additional Details : Gewicht : 0.00010kg
Description
This gene encodes a member of the adenylate kinase family of enzymes. The encoded protein is localized to the mitochondrial matrix. Adenylate kinases regulate the adenine and guanine nucleotide compositions within a cell by catalyzing the reversible transfer of phosphate group among these nucleotides. Five isozymes of adenylate kinase have been identified in vertebrates. Expression of these isozymes is tissue-specific and developmentally regulated. A pseudogene for this gene has been located on chromosome 17. Three transcript variants encoding the same protein have been identified for this gene. Sequence alignment suggests that the gene defined by NM_013410, NM_203464, and NM_001005353 is located on chromosome 1. [provided by RefSeq]
Sequence: MASKLLRAVILGPPGSGKGTVCQRIAQNFGLQHLSSGHFLRENIKASTEVGEMAKQYIEKSLLVPDHVITRLMMSELENRRGQHWLLDGFPRTLGQAEALDKICEVDLVISLNIPFETLKDRLSRRWIHPPSGRVYNLDFNPPHVHGIDDVTGEPLVQQEDDKPEAVAARLRQYKDVAKPVIELYKSRGVLHQFSGTETNKIWPYVYTLFSNKITPIQSKEAYSpecifications
NP_001002921.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
51.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
RUO | |
AK3L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
205 | |
AK3L1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AK3/AK4/MGC166959 | |
AK3L1 | |
Recombinant | |
wheat germ expression system |