Learn More
Abnova™ Human ADAMTSL4 Full-length ORF (AAH27478.2, 1 a.a. - 424 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00054507-P02.10ug
Additional Details : Gewicht : 0.00010kg
Description
This gene is a member of ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs)-like gene family and encodes a protein with protein with seven thrombospondin type 1 repeats. The thrombospondin type 1 repeat domain is found in many proteins with diverse biological functions including cellular adhesion, angiogenesis, and patterning of the developing nervous system. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq]
Sequence: MPAPPHPRTPLGSPAAYWKRVGHSACSASCGKGVWRPIFLCISRESGEELDERSCAAGARPPASPEPCHGTPCPPYWEAGEWTSCSRSCGPGTQHRQLQCRQEFGGGGSSVPPERCGHLPRPNITQSCQLRLCGHWEVGSPWSQCSVRCGRGQRSRQVRCVGNNGDEVSEQECASGPPQPPSREACDMGPCTTAWFHSDWSSKCSAECGTGIQRRSVVCLGSGAALGPGQGEAGAGTGQSCPTGSRPPDMRACSLGPCERTWRWYTGPWGECSSECGSGTQRRDIICVSKLGTEFNVTSPSNCSHLPRPPALQPCQGQACQDRWFSTPWSPCSRSCQGGTQTREVQCLSTNQTLSTRCPPQLRPSRKRPCNSQPCSQRPDDQCKDSSPHCPLVVQARLCVYPYYTATCCRSCAHVLERSPQDPSSpecifications
AAH27478.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.43kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ADAMTSL4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
54507 | |
ADAMTSL4 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TSRC1 | |
ADAMTSL4 | |
Recombinant | |
wheat germ expression system |