missing translation for 'onlineSavingsMsg'
Learn More
Abnova™ Human ACTN4 Full-length ORF (NP_004915.2, 1 a.a. - 911 a.a.) Recombinant Protein with GST-tag at N-terminal Artikelnummer.: 16131231
Verfügbar ab: 01-09-2025
Nur noch null auf Lager
Zum Warenkorb hinzufügen

Abnova™ Human ACTN4 Full-length ORF (NP_004915.2, 1 a.a. - 911 a.a.) Recombinant Protein with GST-tag at N-terminal

Artikelnummer. 16131231
10 μg, 10 Mikrogramm
Click to view available options
Menge:
10 μg
25 μg
Packungsgröße:
10 Mikrogramm
25 Mikrogramm
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16131231

Marke: Abnova™ H00000081P01.10ug

Nur noch null auf Lager
Verfügbar ab: 01-09-2025
Zum Warenkorb hinzufügen
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Used for AP, Array, ELISA, WB-Re

Alpha actinins belong to the spectrin gene superfamily which represents a diverse group of cytoskeletal proteins, including the alpha and beta spectrins and dystrophins. Alpha actinin is an actin-binding protein with multiple roles in different cell types. In nonmuscle cells, the cytoskeletal isoform is found along microfilament bundles and adherens-type junctions, where it is involved in binding actin to the membrane. In contrast, skeletal, cardiac, and smooth muscle isoforms are localized to the Z-disc and analogous dense bodies, where they help anchor the myofibrillar actin filaments. This gene encodes a nonmuscle, alpha actinin isoform which is concentrated in the cytoplasm, and thought to be involved in metastatic processes. Mutations in this gene have been associated with focal and segmental glomerulosclerosis. [provided by RefSeq]

Sequence: MVDYHAANQSYQYGPSSAGNGAGGGGSMGDYMAQEDDWDRDLLLDPAWEKQQRKTFTAWCNSHLRKAGTQIENIDEDFRDGLKLMLLLEVISGERLPKPERGKMRVHKINNVNKALDFIASKGVKLVSIGAEEIVDGNAKMTLGMIWTIILRFAIQDISVEETSAKEGLLLWCQRKTAPYKNVNVQNFHISWKDGLAFNALIHRHRPELIEYDKLRKDDPVTNLNNAFEVAEKYLDIPKMLDAEDIVNTARPDEKAIMTYVSSFYHAFSGAQKAETAANRICKVLAVNQENEHLMEDYEKLASDLLEWIRRTIPWLEDRVPQKTIQEMQQKLEDFRDYRRVHKPPKVQEKCQLEINFNTLQTKLRLSNRPAFMPSEGKMVSDINNGWQHLEQAEKGYEEWLLNEIRRLERLDHLAEKFRQKASIHEAWTDGKEAMLKHRDYETATLSDIKALIRKHEAFESDLAAHQDRVEQIAAIAQELNELDYYDSHNVNTRCQKICDQWDALGSLTHSRREALEKTEKQLEAIDQLHLEYAKRAAPFNNWMESAMEDLQDMFIVHTIEEIEGLISAHDQFKSTLPDADREREAILAIHKEAQRIAESNHIKLSGSNPYTTVTPQIINSKWEKVQQLVPKRDHALLEEQSKQQSNEHLRRQFASQANVVGPWIQTKMEEIGRISIEMNGTLEDQLSHLKQYERSIVDYKPNLDLLEQQHQLIQEALIFDNKHTNYTMEHIRVGWEQLLTTIARTINEVENQILTRDAKGISQEQMQEFRASFNHFDKDHGGALGPEEFKACLISLGYDVENDRQGEAEFNRIMSLVDPNHSGLVTFQAFIDFMSRETTDTDTADQVIASFKVLAGDKNFITAEELRRELPPDQAEYCIARMAPYQGPDAVPGALDYKSFSTALYGESDL

Spezifikation

Zugriffsnummer NP_004915.2
Zur Verwendung mit (Anwendung) Antibody Production, Protein Array, ELISA, Western Blot
Zusammensetzung 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gen-ID (Entrez) 81
Molekulargewicht 131.3kDa
Name ACTN4 (Human) Recombinant Protein (P01)
Reinigungsverfahren Glutathione Sepharose 4 Fast Flow
Qualitätskontrollen 12.5% SDS-PAGE Stained with Coomassie Blue.
Menge 10 μg
Lagerungsbedingungen Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Kennzeichnung RUO
Gen-Alias ACTININ-4/DKFZp686K23158/FSGS/FSGS1
Gebräuchliche Bezeichnung ACTN4
Gensymbol ACTN4
Spezies Wheat Germ (in vitro)
Rekombinant Recombinant
Proteinmarkierung GST
Expressionssystem wheat germ expression system
Form Liquid
Mehr anzeigen Weniger anzeigen
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Abnova™ Human ACTN4 Full-length ORF (NP_004915.2, 1 a.a. - 911 a.a.) Recombinant Protein with GST-tag at N-terminal >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt