Learn More
Abnova™ Human ACO1 Full-length ORF (NP_002188.1, 1 a.a. - 889 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marke: Abnova™ H00000048-P02.10ug
Additional Details : Gewicht : 0.00010kg
Beschreibung
Aconitase 1, also known as iron regulatory element binding protein 1 (IREB1), is a cytosolic protein which binds to iron-responsive elements (IREs). IREs are stem-loop structures found in the 5' UTR of ferritin mRNA, and in the 3' UTR of transferrin receptor mRNA. The iron-induced binding to the IRE results in repression of translation of ferritin mRNA, and inhibition of degradation of the otherwise rapidly degrading transferrin receptor mRNA. Thus, IREB1 plays a central role in cellular iron homeostasis. It was also shown to have aconitase activity, and hence grouped with the aconitase family of enzymes. [provided by RefSeq]
Sequence: MSNPFAHLAEPLDPVQPGKKFFNLNKLEDSRYGRLPFSIRVLLEAAIRNCDEFLVKKQDIENILHWNVTQHKNIEVPFKPARVILQDFTGVPAVVDFAAMRDAVKKLGGDPEKINPVCPADLVIDHSIQVDFNRRADSLQKNQDLEFERNRERFEFLKWGSQAFHNMRIIPPGSGIIHQVNLEYLARVVFDQDGYYYPDSLVGTDSHTTMIDGLGILGWGVGGIEAEAVMLGQPISMVLPQVIGYRLMGKPHPLVTSTDIVLTITKHLRQVGVVGKFVEFFGPGVAQLSIADRATIANMCPEYGATAAFFPVDEVSITYLVQTGRDEEKLKYIKKYLQAVGMFRDFNDPSQDPDFTQVVELDLKTVVPCCSGPKRPQDKVAVSDMKKDFESCLGAKQGFKGFQVAPEHHNDHKTFIYDNTEFTLAHGSVVIAAITSCTNTSNPSVMLGAGLLAKKAVDAGLNVMPYIKTSLSPGSGVVTYYLQESGVMPYLSQLGFDVVGYGCMTCIGNSGPLPEPVVEAITQGDLVAVGVLSGNRNFEGRVHPNTRANYLASPPLVIAYAIAGTIRIDFEKEPLGVNAKGQQVFLKDIWPTRDEIQAVERQYVIPGMFKEVYQKIETVNESWNALATPSDKLFFWNSKSTYIKSPPFFENLTLDLQPPKSIVDAYVLLNLGDSVTTDHISPAGNIARNSPAARYLTNRGLTPREFNSYGSRRGNDAVMARGTFANIRLLNRFLNKQAPQTIHLPSGEILDVFDAAERYQQAGLPLIVLAGKEYGAGSSRDWAAKGPFLLGIKAVLAESYERIHRSNLVGMGVIPLEYLPGENADALGLTGQERYTIIIPENLKPQMKVQVKLDTGKTFQAVMRFDTDVELTYFLNGGILNYMIRKMAKSpezifikation
NP_002188.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
124.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
RUO | |
ACO1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
48 | |
ACO1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ACONS/IREB1/IREBP/IREBP1/IRP1 | |
ACO1 | |
Recombinant | |
wheat germ expression system |