missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HTR2B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533304
Additional Details : Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Orangutan, Gibbon, Monkey, Marmoset (100%) Bovine, Rabbit, Pig (88%).
HTR2B Polyclonal antibody specifically detects HTR2B in Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
HTR2B | |
Polyclonal | |
Unconjugated | |
HTR2B | |
5-HT 2B receptor; 5-HT(2B); 5-HT2B; 5-HT-2B; 5HT2B Receptor; 5-HT2b receptor; 5-HT-2F; 5-hydroxytryptamine (serotonin) receptor 2B; 5-hydroxytryptamine (serotonin) receptor 2B, G protein-coupled; 5-hydroxytryptamine 2B receptor; 5-hydroxytryptamine receptor 2B; 5-hydroxytryptamine receptor 2B variant b; AJ012488; AV377389; HTR2B; NP75 protein; serotonin receptor 2B; Srl; Stomach fundus serotonin receptor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15559, 29581, 3357 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.71 mg/mL | |
PBS with 0.1% sodium azide | |
P30994, P41595, Q02152 | |
HTR2B | |
Synthetic 17 amino acid peptide from 3rd cytoplasmic domain of human 5HT2B Receptor. | |
50 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |