missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hsc70 Mouse anti-Human, Mouse, Rat, Clone: 1A4B3, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66442-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
The product encoded by this gene belongs to the heat shock protein 70 family which contains both heat-inducible and constitutively expressed members. The latter are called heat-shock cognate proteins. This gene encodes a heat-shock cognate protein. This protein binds to nascent polypeptides to facilitate correct folding. It also functions as an ATPase in the disassembly of clathrin-coated vesicles during transport of membrane components through the cell. Two alternatively spliced variants have been characterized to date.Especificaciones
Hsc70 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P11142, P63017, P63018 | |
HSPA8 | |
Hsc70 Fusion Protein Ag17910 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
1A4B3 | |
Unconjugated | |
HSPA8 | |
Heat shock 70 kDa protein 8, heat shock 70kDa protein 8, HSC54, Hsc70, HSC71, HSP71, HSP73, HSPA10, HSPA8, LAP1, NIP71 | |
Mouse | |
Protein G | |
RUO | |
15481, 24468, 3312 | |
-20°C | |
Liquid |