missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRSP12 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | HRSP12 |
---|---|
Konzentration | 0.27 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850894
|
Proteintech
12930-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840894
|
Proteintech
12930-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 14-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Product No. 12930-1-AP is a rabbit polyclonal antibody raised from human full-length UK114.
Endoribonuclease responsible for the inhibition of the translation by cleaving mRNA. Inhibits cell-free protein synthesis. Cleaves phosphodiester bonds only in single-stranded RNA.Spezifikation
HRSP12 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
P52758, P52759 | |
10247, 65151 | |
HRSP12 Fusion Protein Ag3628 | |
Primary | |
-20°C | |
RIDA |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
heat responsive protein 12, HRSP12, P14.5, PSP, Ribonuclease UK114, UK114, UK114 antigen homolog | |
Rida | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |