missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HRPT2/CDC73 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 4B4H2, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66490-1-IG-20UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
U2 auxiliary factor (U2AF), comprised of a large and a small subunit, is a non-snRNP protein required for the binding of U2 snRNP to the pre-mRNA branch site. This gene encodes the U2AF large subunit which contains a sequence-specific RNA-binding region with 3 RNA recognition motifs and an Arg/Ser-rich domain necessary for splicing. The large subunit binds to the polypyrimidine tract of introns early during spliceosome assembly. Multiple transcript variants have been detected for this gene, but the full-length natures of only two have been determined to date.Spezifikation
HRPT2/CDC73 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4V8C8, Q6P1J9, Q8JZM7 | |
CDC73 | |
HRPT2; CDC73 Fusion Protein ag2959 | |
20 μL | |
Primary | |
Human, Rat, Porcine, Mouse | |
Antibody | |
IgG2a |
Western Blot, Immunohistochemistry (Paraffin) | |
4B4H2 | |
Unconjugated | |
CDC73 | |
C1orf28, CDC73, HPT JT, HRPT2, Hyperparathyroidism 2 protein, Parafibromin | |
Mouse | |
Protein A | |
RUO | |
100526068, 214498, 304832, 79577 | |
-20°C | |
Liquid |